Learn More
IGFBP-4 Mouse, HRP, Clone: IBP185, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP232772H
Catalog No. NBP232772H
Add to cart
Description
Description
IGFBP-4 Monoclonal antibody specifically detects Antigen in Human, Mouse samples. It is validated for Enzyme Linked Immunosorbent Assay.
Specifications
Specifications
IGFBP-4
IBP185
ELISA
Purified
IGFBP4
Human C-terminal IGFBP-4 fragment (CT-IGFBP-4)
Protein A purified
RUO
Primary
3487
ELISA
HRP
PBS with No Preservative
BP-4, HT29-IGFBP, IBP4insulin-like growth factor-binding protein 4, IGF-binding protein 4, IGFBP-4IBP-4, insulin-like growth factor binding protein 4
Mouse
IgG2b
0.2 ml
Store at 4C in the dark.
Monoclonal
Human, Mouse