Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IGFBP-5 Rat anti-Mouse, DyLight 488, Clone: RM0097-12C23, Novus Biologicals
Rat Monoclonal Antibody
Manufacturer: Novus Biologicals NBP122453G
Description
IGFBP-5 Monoclonal antibody specifically detects IGFBP-5 in Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
IGFBP-5 | |
RM0097-12C23 | |
Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin | |
Purified | |
IGFBP5 | |
Mouse recombinant protein of IGFBP-5 | |
Protein G purified | |
Store at 4C in the dark. | |
Monoclonal | |
This antibody detects mouse IGFBP-5 in Western blotting. No cross activity to other mouse IGFBPs. |
Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot | |
DyLight 488 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
IBP5IBP-5, IGF-binding protein 5, IGFBP-5, insulin-like growth factor binding protein 5, insulin-like growth factor-binding protein 5 | |
Rat | |
IgG2 | |
0.1 ml | |
Primary | |
3488 | |
Mouse |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title