Learn More
IGFBP-rp1/IGFBP-7 Mouse anti-Human, Alexa Fluor™ 532, Clone: MM0346-3N37, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP212368AF532
Catalog No. NP212368A32
Add to cart
Description
Description
IGFBP-rp1/IGFBP-7 Monoclonal antibody specifically detects IGFBP-rp1/IGFBP-7 in Human samples. It is validated for Western Blot.
Specifications
Specifications
IGFBP-rp1/IGFBP-7
MM0346-3N37
Western Blot
angiomodulin, FSTL2, IBP-7, IGFBP-7PGI2-stimulating factor, IGFBP-7v, IGFBP-rP1, IGFBPRP1, insulin-like growth factor binding protein 7, insulin-like growth factor-binding protein 7, MAC25 protein, MAC25IGF-binding protein 7, Prostacyclin-stimulating factor, PSFAGM, TAF, Tumor-derived adhesion factor
Mouse
IgG1
Store at 4°C in the dark.
Monoclonal
Human
Western Blot
Alexa Fluor™ 532
50 mM sodium borate with 0.05% sodium azide
IGFBP7
Human IGFBP7
0.1 mL
Primary
3490