Learn More
IGFBP-rp1/IGFBP-7 Mouse anti-Human, DyLight 350, Clone: 1D9E7, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP237313UV
Catalog No. NBP237313UV
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Specifications
Specifications
IGFBP-rp1/IGFBP-7
1D9E7
Western Blot, ELISA, Immunohistochemistry
Purified
IGFBP7
Purified recombinant fragment of human IGFBP7 (AA: 52-156) expressed in E. Coli.
Protein A or G purified
Biologically Active Proteins
Primary
3490
ELISA, Immunohistochemistry, Western Blot
DyLight 350
50mM Sodium Borate with 0.05% Sodium Azide
angiomodulin, FSTL2, IBP-7, IGFBP-7PGI2-stimulating factor, IGFBP-7v, IGFBP-rP1, IGFBPRP1, insulin-like growth factor binding protein 7, insulin-like growth factor-binding protein 7, MAC25 protein, MAC25IGF-binding protein 7, Prostacyclin-stimulating factor, PSFAGM, TAF, Tumor-derived adhesion factor
Mouse
IgG1
0.1 ml
Store at 4C in the dark.
Monoclonal
Human