Learn More
Aviva Systems Biology Corp IGFBP2 Antibody - middle region (ARP54334_P050) | 100uL
Supplier:  Aviva Systems Biology Corp ARP54334_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against IGFBP2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human IGFBP2. Alternate names: IBP2, IGF-BP53ApplicationWB. Species reactivity: Cow, Dog, Goat, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Sheep
Catalog No. 50-014-61178