Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IGFBP3 Mouse anti-Human, Clone: OTI3B9, lyophilized, TrueMAB™
Mouse Monoclonal Antibody
Supplier: OriGene CF807469
Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
Insulin like growth factor binding protein 3 (IGFBP3) is a member of the superfamily of insulin like growth factor (IGF) binding proteins which include six high affinity IGF binding proteins (IGFBP) and at least four lowaffinity binding proteins referred to as IGFBP related proteins (IGFBPrP). The IGFBP members are cysteine rich proteins with conserved cysteine residues clustered in the amino terminal and the carboxy terminal regions of the molecule. The cDNA sequence encoding the mature human IGFBP3 is fused to the signal peptide of CD33. The 265 amino acid residue recombit mature human IGFBP3 has a calculated molecular mass of approximately 29 kDa. Due to glycosylation, the recombit protein migrates as 41 kDa in SDS-PAGE.Specifications
IGFBP3 | |
Monoclonal | |
Unconjugated | |
IGFBP3 | |
BP-53, IBP3 | |
Mouse | |
Affinity Chromatography | |
RUO | |
3486 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Western Blot | |
OTI3B9 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
P17936 | |
IGFBP3 | |
Human recombit protein fragment corresponding to amino acids 28-297 of human IGFBP3 produced in E.coli. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG2b |
Provide Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction