Learn More
Invitrogen™ IGFBP4 Human ELISA Kit
Sandwich ELISA Kit
$596.00 - $2930.00
Specifications
Accession Number | P22692 |
---|---|
Assay Sensitivity | 250 pg/mL |
Assay | Solid-phase sandwich ELISA |
Conjugate | Biotin |
Product Type | ELISA Kit |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
EHIGFBP4
|
Invitrogen™
EHIGFBP4 |
96 Tests |
Pack for $596.00
|
|
|||||
EHIGFBP4X5
|
Invitrogen™
EHIGFBP4X5 |
5 x 96 Tests |
Each for $2,930.00
|
|
|||||
Description
The Human Insulin-like Growth Factor Binding Protein 4 (Hu IGFBP4) ELISA quantitates Hu IGFBP4 in human serum, plasma, or cell culture medium. The assay will exclusively recognize both natural and recombinant Hu IGFBP4. Principle of the method The Human IGFBP4 solid-phase sandwich ELISA (enzyme-linked immunosorbent assay) is designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody has been pre-coated in the wells of the supplied microplate. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. The sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen. Rigorous validation Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.
This gene is a member of the insulin-like growth factor binding protein (IGFBP) family and encodes a protein with an IGFBP domain and a thyroglobulin type-I domain. The protein binds both insulin-like growth factors (IGFs) I and II and circulates in the plasma in both glycosylated and non-glycosylated forms. Binding of this protein prolongs the half-life of the IGFs and alters their interaction with cell surface receptors.Specifications
P22692 | |
Solid-phase sandwich ELISA | |
ELISA Kit | |
Plasma, Serum, Supernatant | |
Human | |
Colorimetric Microplate Reader | |
BP-4,HT29-IGFBP,IBP4,IGFBP-4 | |
<10% | |
RUO | |
2°C to 8°C | |
Human | |
4 hr. 45 min. |
250 pg/mL | |
Biotin | |
Growth Factors & Receptors | |
This ELISA kit shows no cross-reactivity with any of the cytokines tested: Human Angiogenin, BDNF, BLC, ENA-78, FGF-4, IGFBP-1, IGFBP-2, IGFBP-3, IL-1 alpha, IL-1 beta, IL-2, IL-3, IL-4, IL-5, IL-7, IL- 8, IL-9, IL-10, IL-11, IL-12 p70, IL-12 p40, IL-13, IL-15, IL-309, IP-10, GCSF, GM-CSF, IFN-gamma, Leptin, MCP-1, MCP-2, MCP-3, MDC, MIP-1 alpha, MIP-1 beta, MIP-1 delta, PARC, PDGF, RANTES, SCF, TARC, TGF-beta, TIMP-1, TIMP-2, TNF-alpha, TNF-beta, TPO, VEGF. | |
ELISA, Protein Assays, Protein Biology, Cancer Biology, Immunology, Neuroscience, Neurobiology, Protein Detection, Protein Analysis | |
3487 | |
<12% | |
HRP | |
Signal Transduction | |
BP-4, HT29-IGFBP, IBP4, IGFBP-4 | |
1 hr. 20 min. |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.