Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IGFLR1/TMEM149 Antibody (905338), HRP, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals FAB8296H
Description
IGFLR1/TMEM149 Monoclonal antibody specifically detects IGFLR1/TMEM149 in Human samples. It is validated for Flow Cytometry, Blocking Assay, CyTOFSpecifications
| IGFLR1/TMEM149 | |
| Monoclonal | |
| HRP | |
| PBS | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry, Blocking Assay, CyTOF | |
| 905338 | |
| Flow Cytometry, Blockade of Receptor-ligand Interaction, CyTOF-ready | |
| FLJ22573, transmembrane protein 149, U2 small nuclear RNA auxiliary factor 1-like 4, U2(RNU2) small nuclear RNA auxiliary factor 1-like 4, U2AF1L4 | |
| Human embryonic kidney cell line HEK293-derived recombinant human IGFLR1, Met1-Pro163, Accession # Q9H665 | |
| 0.1 mL | |
| Signal Transduction | |
| 79713 | |
| Store at 4C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction