Learn More
IGF-I R/IGF1R Human anti-Human, Alexa Fluor 405, Clone: Hu14, R&D Systems
Human Monoclonal Antibody
Manufacturer:  R&D Systems FAB9718V100UG
Catalog No. FAB9718V-100
Due to product restrictions, please to purchase or view availability for this product.
This item is not returnable. View return policy
Description
Description
IGF-I R/IGF1R Monoclonal antibody specifically detects IGF-I R/IGF1R in Human samples. It is validated for Flow Cytometry.
Specifications
Specifications
IGF-I R/IGF1R
Hu14
Flow Cytometry 0.25-1μg/106 cells
Supernatant
Human
IgG1
100μg
Primary
3480
Human
Flow Cytometry
Alexa Fluor 405
Supplied 0.2Mg/mL in a saline solution containing BSA and Sodium Azide.
CD221, CD221 antigen, EC 2.7.10, EC 2.7.10.1, IGF1R, IGF-1R, IGF-I receptor, IGFIR, IGF-IR, IGFR, insulin-like growth factor 1 receptor, Insulin-like growth factor I receptor, JTK13, MGC142170, MGC142172, MGC18216, soluble IGF1R variant 1, soluble IGF1R variant 2
Human IGF-I R/IGF1R
Protein A or G purified from cell culture supernatant
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied.
Monoclonal
Detects human IGF-I R/IGF1R based on Teprotumumab therapeutic antibody. This non-therapeutic antibody uses the same variable region sequence as the therapeutic antibody Teprotumumab. This product is for research use only.