Learn More
IGFLR1/TMEM149 Mouse anti-Human, Alexa Fluor 594, Clone: 905338, R&D Systems™
Mouse Monoclonal Antibody
Manufacturer:  R&D Systems FAB8296T100UG
Catalog No. FAB8296T100
Add to cart
This item is not returnable. View return policy
Description
Description
IGFLR1/TMEM149 Monoclonal specifically detects IGFLR1/TMEM149 in Human samples. It is validated for Flow Cytometry.
Specifications
Specifications
IGFLR1/TMEM149
Monoclonal
0.2 mg/mL
Flow Cytometry 0.25-1 ug/10^6 cells
Q9H665
IGFLR1
Human embryonic kidney cell line HEK293-derived recombinant human IGFLR1 Met1-Pro163 Accession # Q9H665
100 ug
Store the unopened product at 2 - 8 degreesC. Do not use past expiration date.
Monoclonal
Detects human IGFLR1in direct ELISAs.
Flow Cytometry
905338
Alexa Fluor 594
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide
FLJ22573, TMEM149, transmembrane protein 149, U2 small nuclear RNA auxiliary factor 1-like 4, U2(RNU2) small nuclear RNA auxiliary factor 1-like 4, U2AF1L4
Mouse
IgG1
RUO
Primary
79713
Human
SDS
Documents
Product Certifications
Promotions
Promotions
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Cancel Submit
For Research Use Only (RUO)