Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IgG, Armenian Hamster, Clone: 299Arm, Isotype Control, Novus Biologicals™

Armenian Hamster Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP259616500UG

Catalog No. NBP59616500


Add to cart

Description

Description

IgG Monoclonal antibody specifically detects IgG in Armenian Hamster samples. It is validated for Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, In vitro assay, Control.
Specifications

Specifications

IgG
299Arm
Flow Cytometry 1:10-1:1000, ELISA 1:100-1:2000, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:10-1:500, In vitro assay, Control
Affinity purified, Liquid
IgG
500 μg
Isotype Control
Armenian Hamster
Control, ELISA, Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, In vitro Assay
Unconjugated
PBS (pH 7.2)
Armenian Hamster
Affinity purified
Store at 4°C. Do not freeze.
Monoclonal
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit