Learn More
F(ab')2-Goat anti-Mouse IgG (H+L) Secondary Antibody, eFluor™ 660, eBioscience™, Invitrogen™
F(ab')2-Goat Polyclonal Secondary Antibody
$201.00
Specifications
Antigen | Mouse IgG (H+L) |
---|---|
Concentration | 0.2 mg/mL |
Classification | Polyclonal |
Conjugate | eFluor 660 |
Form | Liquid |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-246-037
|
Invitrogen
50401082 |
100 μg |
Each of 1 for $201.00
|
|
|||||
Description
Description: The goat anti-mouse IgG polyclonal antibody reacts with the heavy and light chains of mouse IgG, with minimal cross reactivity to human, rat, bovine, goat, hamster, and rabbit serum proteins. Reactivity has been shown to IgG1, IgG2a, IgG2b, and IgG3, as well as some reactivity to IgM. The antibody is a F(ab')2 fragment which contains the 2 Fab domains still linked through the hinge region of the immunoglobulin. Removal of the Fc region helps to reduce issues typically found with Fc-mediated binding. This polyclonal has been validated as a secondary reagent (its ability to recognize cells stained with a mouse IgG followed by this polyclonal antibody). Applications Reported: This polyclonal antibody has been reported for use in flow cytometric analysis, immunohistochemistry and immunocytochemistry. Applications Tested: This polyclonal antibody has been tested by flow cytometric analysis of normal human peripheral blood cells stained first with a mouse anti-human antibody. This can be used at less than or equal to 0.5 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. This polyclonal antibody has been tested by immunohistochemistry on formalin-fixed paraffin embedded sections that have been stained first with a mouse anti-human antibody.
Mouse IgG (H+L) Polyclonal antibody specifically detects Mouse IgG (H+L) in Mouse samples. It is validated for Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Paraffin).Specifications
Mouse IgG (H+L) | |
Polyclonal | |
Liquid | |
RUO | |
Ig | |
Affinity chromatography |
0.2 mg/mL | |
eFluor 660 | |
Goat | |
Mouse | |
Secondary | |
4° C, store in dark, DO NOT FREEZE! |
For Research Use Only.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.