Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IgG F(ab)2, Guinea Pig, HRP, Polyclonal, Isotype Control, Novus Biologicals™

Guinea Pig Polyclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP1971560.5MG

Catalog No. NBP19715605


Add to cart

Description

Description

IgG F(ab)2 Polyclonal antibody specifically detects IgG F(ab)2 in Guinea Pig samples. It is validated for Western Blot, ELISA.
Specifications

Specifications

IgG F(ab)2
HRP
0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2, 10 mg/mL (BSA) - Immunoglobulin and Protease free
Guinea Pig
Ion exchange chromatography
Store lyophilized antibody at 4°C in the dark. Aliquot reconstituted liquid and store at -20°C. Avoid freeze-thaw cycles.
Polyclonal
Reconstitute with 1.0 mL deionized water.
ELISA, Western Blot
Western Blot, ELISA
Conjugated, Lyophilized
IgG
0.5 mg
Isotype Control
Assay by immunoelectrophoresis resulted in a single precipitin arc against anti-Peroxidase, anti-Guinea Pig IgG, anti-Guinea Pig IgG F(ab)2 and anti-Guinea Pig Serum. No reaction was observed against anti-Guinea Pig IgG F(c) or anti-Pepsin.
Guinea Pig
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit