Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IgG F(ab)2, Rat, Agarose, Polyclonal, Isotype Control, Novus Biologicals™

Rat Polyclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP1972011MG

Catalog No. NBP1972011


Add to cart

Description

Description

IgG F(ab)2 Polyclonal antibody specifically detects IgG F(ab)2 in Rat samples. It is validated for Immunoprecipitation, SDS-Page.
Specifications

Specifications

IgG F(ab)2
Agarose
0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2
Rat
Multi-step
Store at 4°C. Do not freeze.
Polyclonal
Immunoprecipitation, Western Blot
Immunoprecipitation 1:10 - 1:500, SDS-Page 1:100 - 1:2000
Conjugated
IgG
1 mg
Isotype Control
Rat
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit