Learn More
IgG F(ab) Goat anti-Human, Biotin, Polyclonal, Novus Biologicals™
Goat Polyclonal Antibody
Manufacturer:  Novus Biologicals NBP2694381000UL
Catalog No. NB002848
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
IgG F(ab) Polyclonal antibody specifically detects IgG F(ab) in Human samples. It is validated for ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Frozen).
Specifications
Specifications
IgG F(ab)
LYOPH
ELISA 1:100-1:2000, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1:10 - 1:500, Immunohistochemistry-Frozen 1:10 - 1:500
Purified
Purified Fab from normal IgG isolated from pooled human serum. Freund's complete adjuvant is used in the first step of the immunization procedure.
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
It is reconstituted by adding 1 mL sterile distilled water, spun down to remove insoluble particles, d, spun down again and use.ivided into small aliquots, frozen and stored at or below -20°C. Prior to use, an aliquot is thawed slowly at ambient temperature
ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Frozen)
Biotin
Lyophilized from PBS (pH 7.2)
Goat
1000 μL
Polyclonal
Human