Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IgG F(ab) Goat anti-Rabbit, Biotin, Clone: RMG01, Secondary Antibody, Novus Biologicals™

Goat Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP262014B

Catalog No. NBP262014B


Add to cart

Description

Description

IgG F(ab) Monoclonal antibody specifically detects IgG F(ab) in Rabbit samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunoprecipitation, Immunocytochemistry.
Specifications

Specifications

IgG F(ab)
RMG01
Biotin
50% Glycerol/PBS with 1% BSA with 0.09% Sodium Azide
Goat
IgG
50 ug
Secondary
This antibody reacts to the Fab region of Rabbit IgG. No cross reactivity with human IgG, rat IgG, or mouse IgG
ELISA, Flow Cytometry, Immunocytochemistry, Immunohistochemistry, Immunoprecipitation, Western Blot
1 mg/mL
Western Blot 0.1ug/mL - 0.5ug/mL, Flow Cytometry, ELISA 0.01ug/mL - 0.5ug/mL, Immunohistochemistry 0.5ug/mL - 2ug/mL, Immunoprecipitation, Immunocytochemistry 0.5ug/mL - 2ug/mL
Purified
Rabbit IgG
Protein G purified
Store at -20°C. Avoid freeze-thaw cycles.
Monoclonal
Rabbit
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit