Learn More
Athens Research And Technology IncSupplier Diversity Partner IGG FAB FRAGMENT HU PLASMA 1MG DFS Item
Manufacturer:  Athens Research And Technology IncSupplier Diversity Partner 1616090707FAB1MG
Immunoglobulin G, Fab Fragment, Human Plasma: 1 MG; Frozen in 20 mM phosphate, pH 7.4, with 150 mM NaCl & 0.05% NaN3; MW 55,000; EC=1.35; Purity: > =95% by SDS-PAGE. No reaction by IEP to antisera to human IgG-fc. Native; Prepared from human plasma.
Catalog No. 50-489-785