Learn More
IgG Fab Goat anti-Rabbit, Clone: RMG01, Abnova™
Goat monoclonal antibody raised against full length native rabbit IgG Fab.
Manufacturer:  Abnova Corporation MAB12773
Catalog No. 89-934-832
Add to cart
Description
Description
Specifications
Specifications
IgG Fab
RMG01
Goat monoclonal antibody raised against full length native rabbit IgG Fab.
In PBS (50% glycerol, 1% BSA, 0.09% sodium azide)
Goat
IgG
100 ug
Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Monoclonal
Rabbit
ELISA, Flow Cytometry, Immunocytochemistry, Immunohistochemistry (PFA fixed), Immunoprecipitation, Western Blot
Unconjugated
ELISA (0.01 ug/mL- 0.5 ug/mL) Flow Cytometry Immunocytochemistry (0.5 ug/mL-2 ug/mL) Immunohistochemistry (0.5 ug/mL-2 ug/mL) Immunoprecipitation Western Blot (0.1 ug/mL-0.5 ug/mL) The optimal working dilution should be determined by the
Liquid
Native purified rabbit IgG.
Protein G purification
RUO
Primary
This antibody reacts to the Fab region of Rabbit IgG. No cross reactivity with human IgG, rat IgG, or mouse IgG