Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IgG Fc, Canine, Biotin, Polyclonal, Isotype Control, Novus Biologicals™

Canine Polyclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP1972370.5MG

Catalog No. NBP19723705


Add to cart

Description

Description

IgG Fc Polyclonal antibody specifically detects IgG Fc in Canine samples. It is validated for Immunocytochemistry/Immunofluorescence, Fluorophore-linked immunosorbent assay.
Specifications

Specifications

IgG Fc
Biotin
0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2, 10 mg/mL (BSA) - Immunoglobulin and Protease free
Canine
Ion exchange chromatography
Store lyophilized antibody at 4°C in the dark. Aliquot reconstituted liquid and store at -20°C. Avoid freeze-thaw cycles.
Polyclonal
Reconstitute with 1.0 mL deionized water.
Immunocytochemistry, Immunofluorescence
Immunocytochemistry/Immunofluorescence, Fluorophore-linked immunosorbent assay
Conjugated, Lyophilized
IgG
0.5 mg
Isotype Control
Assay by immunoelectrophoresis resulted in a single precipitin arc against anti-biotin, anti-Canine IgG, anti-Canine IgG F(c) and anti-Canine Serum. No reaction was observed against anti-Canine IgG F(ab)2 or anti-Papain.
Canine
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit