Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IgG Fc fragment Goat anti-Rat, Biotin, Polyclonal, Secondary Antibody, Novus Biologicals™

Goat Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP261920B

Catalog No. NBP261920B


Add to cart

Description

Description

IgG Fc fragment Polyclonal antibody specifically detects IgG Fc fragment in Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry.
Specifications

Specifications

IgG Fc fragment
0.5 mg/mL
Western Blot 1:10000 - 1:200000, ELISA 1:10000 - 1:200000, Immunohistochemistry 1:250 - 1:2500, Immunocytochemistry 1:100 - 1:500
Affinity Purified
Rat IgG-Fc Fragment
Immunogen affinity purified
Adaptive Immunity
Secondary
Antiserum was solid phase adsorbed to ensure class specificity. Antiserum was cross adsorbed using human and mouse immunosorbents to remove cross reactive antibodies. The antibody to rat IgG was isolated by affinity chromatography using antigen coupled to agarose beads. F(ab')2 fragments were generated using a pepsin digestion. Fc fragments and whole IgG molecules have been removed. Fragments were conjugated to biotin. Antibody concentration was determined by extinction coefficient: absorbance at 280 nm of 1.4 equals 1.0 mg of IgG. By immunoelectrophoresis and ELISA this antibody reacts specifically with rat IgG. Cross reactivity with IgA and IgM is negligible. No antibody was detected against non-immunoglobulin serum proteins. Less than 1% cross reactivity to human and mouse IgG was detected. This antibody may cross react with IgG from other species.
ELISA, Immunocytochemistry, Immunohistochemistry, Western Blot
Biotin
PBS containing 0.2% BSA with 0.09% Sodium Azide
Goat
IgG
1 ml
Store at 4C. Do not freeze.
Polyclonal
Rat
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit