Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IgG, Goat, Unconjugated, Polyclonal, Isotype Control, Novus Biologicals™

Goat Polyclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP19705710MG

Catalog No. NBP19705710


Add to cart

Description

Description

IgG Polyclonal antibody specifically detects IgG in Goat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, SDS-Page.
Specifications

Specifications

IgG
Unconjugated
0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2
Goat
Ion exchange chromatography
Store lyophilized antibody at 4°C. Aliquot reconstituted liquid and store at -20°C. Avoid freeze-thaw cycles.
Polyclonal
Reconstitute with 1.0 mL deionized water.
ELISA, Immunohistochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot
Western Blot 1:100 - 1:2000, ELISA 1:100 - 1:2000, Immunohistochemistry 1:10 - 1:500, Immunohistochemistry-Paraffin 1:10 - 1:500, Immunohistochemistry-Frozen, SDS-Page 1:100 - 1:2000
Lyophilized
IgG
10 mg
Isotype Control
Goat IgG whole molecule assayed by immunoelectrophoresis resulted in a single precipitin arc against anti-Goat IgG and anti-Goat Serum.
Goat
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit