Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Syrian Hamster IgG, Biotin, Isotype Control, eBioscience™ Non-distribution product as customer accommodation.

Syrian Hamster Isotype Control

$84.75 - $173.00

Specifications

Antigen Syrian Hamster IgG
Target Species Not Applicable
Conjugate Biotin
Isotype IgG
Applications Control, Flow Cytometry, Immunohistochemistry
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
50-118-78 Non-distribution product as customer accommodation.
View Documents
Invitrogen
13491481
50μg
Each for $84.75
Add to cart
 
50-118-79 Non-distribution product as customer accommodation.
View Documents
Invitrogen
13491485
500μg
Each for $173.00
Add to cart
 
Description

Description

The isotype of a primary antibody and the application it is being used in can result in background staining. Primary antibody background noise can be caused by binding to Fc receptors on target cells; by non-specific interactions with cellular proteins, carbohydrates, and lipids; or by cell autofluorescence. Isotype control antibodies can act as negative controls to help differentiate non-specific background signal from specific antibody signal because they have no relevant specificity to a target antigen. While isotype controls are most commonly used in flow cytometry, they are useful in other applications such as chromatin immunoprecipitation (ChIP), immunohistochemistry, and gel shifts. Isotype controls should match with the primary antibody species and isotype so that the level of specific staining by the primary antibody may be accurately determined. If using directly labeled primary antibodies, the isotype control works best if conjugated with the same label as the test antibody.

Description: This hamster IgG fraction was purified by affinity chromatography from normal Golden Syrian hamster serum.Applications Reported: Golden Syrian hamster IgG isotype control has been reported for use in surface and intracellular flow cytometric analysis, and immunohistochemical staining.Applications Tested: This Golden Syrian hamster IgG isotype control has been tested by flow cytometric analysis of mouse splenocyte suspension. Isotype control should be used at the same concentration as experimental antibody.Filtration: 0.2µm post-manufacturing filtered.
Specifications

Specifications

Syrian Hamster IgG
Biotin
Control, Flow Cytometry, Immunohistochemistry
Liquid
Isotype Control
Store at 2-8°C. Do not freeze.
Not Applicable
IgG
Polyclonal
Control
PBS with 0.1% gelatin and 0.09% sodium azide; pH 7.2
RUO
SDS

For Research Use Only.

Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit