Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IgG (H+L) Goat anti-Rabbit, AP, Polyclonal, Novus Biologicals™

Goat Polyclonal Antibody

$171.35

Specifications

Antigen IgG (H+L)
Immunogen Purified Rabbit IgG (H&L)
Isotype IgG
Monoclonal or Polyclonal Polyclonal
Primary or Secondary Secondary
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP252984 View Documents Novus Biologicals
NBP252984
0.5 mg Each for $171.35
Add to cart
 
NBP252985 View Documents Novus Biologicals
NBP252985
0.5 mg This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
Description

Description

IgG (H+L) Polyclonal antibody specifically detects IgG (H+L) in Rabbit samples. It is validated for Western Blot, ELISA, Immunohistochemistry.
Specifications

Specifications

IgG (H+L)
IgG
Secondary
Based on Immunoelectrophoresis, this antibody reacts with heavy chains on rabbit IgG and light chains on all rabbit immunoglobulins.
Affinity Purified
0.5 mg
Rabbit
Purified Rabbit IgG (H&L)
Polyclonal
Immunogen affinity purified
AP
Goat
RUO

For Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit