Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IgG (H+L) Rabbit anti-Mouse, Biotin, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

$164.45

Specifications

Antigen IgG (H+L)
Dilution Immunoassay
Formulation 30 mM Triethanolamine, pH 7.2, 5 mM Magnesium Chloride, 0.1 mM Zinc Chloride, 1 % (w/v) BSA, Protease/IgG free with 0.05% Sodium Azide
Immunogen Purified Mouse IgG (H&L)
Monoclonal or Polyclonal Polyclonal
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP252988 View Documents Novus Biologicals
NBP252988
1 mg Each for $164.45
Add to cart
 
Description

Description

IgG (H+L) Polyclonal antibody specifically detects IgG (H+L) in Mouse samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunoassay.
Specifications

Specifications

IgG (H+L)
30 mM Triethanolamine, pH 7.2, 5 mM Magnesium Chloride, 0.1 mM Zinc Chloride, 1 % (w/v) BSA, Protease/IgG free with 0.05% Sodium Azide
Polyclonal
>95%, by SDS-PAGE
Immunoassay
Affinity Purified
1 mg
Mouse
Immunoassay
Purified Mouse IgG (H&L)
Secondary
Based on IEP, this antibody reacts with heavy gamma chains on mouse IgG and light chains on all mouse immunoglobulins
Biotin
Rabbit
RUO

For Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit