Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IgG (H+L) Rabbit anti-Mouse, HRP, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

$228.85

Specifications

Antigen IgG (H+L)
Concentration LYOPH
Dilution Immunomicroscopy 1:20 - 1:2000
Formulation 10 mM Sodium Phosphate, 0.15 M Sodium Chloride, pH 7.2, 1 % (w/v) BSA, Protease/IgG free with 0.1% Kathon
Immunogen Purified Mouse IgG (H&L)
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP252989 View Documents Novus Biologicals
NBP252989
1 mg Each for $228.85
Add to cart
 
Description

Description

IgG (H+L) Polyclonal antibody specifically detects IgG (H+L) in Mouse samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunomicroscopy, Chemiluminescence Immunoassay.
Specifications

Specifications

IgG (H+L)
Immunomicroscopy 1:20 - 1:2000
Purified Mouse IgG (H&L)
Secondary
Rehydrate with 1.1 ml of deionized water and let stand 30 minutes at room temperature to dissolve. (Product has been overfilled to ensure complete recovery.) Centrifuge to remove any particulates. Prepare fresh working dilution daily.
Immunomicroscopy
Affinity Purified
1 mg
Mouse
LYOPH
10 mM Sodium Phosphate, 0.15 M Sodium Chloride, pH 7.2, 1 % (w/v) BSA, Protease/IgG free with 0.1% Kathon
Polyclonal
>95%, by SDS-PAGE
Based on IEP, this antibody reacts with heavy gamma chains on mouse IgG and light chains on all mouse immunoglobulins
HRP
Rabbit
RUO

For Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit