Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IgG Heavy Chain Mouse anti-Human, Biotin, Clone: IG217 + IG266, Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP314095B
This item is not returnable.
View return policy
Description
IgG Heavy Chain Monoclonal antibody specifically detects IgG Heavy Chain in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)Specifications
IgG Heavy Chain | |
Monoclonal | |
Biotin | |
Immunoglobulin G, ImmunoglobulinG | |
Purified human IgG | |
0.1 mL | |
Immunology | |
Human | |
Purified |
Flow Cytometry, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
IG217 + IG266 | |
PBS | |
Mouse | |
Protein A or G purified | |
RUO | |
Primary | |
Store at 4°C in the dark. | |
IgG1 κ, IgG2a κ |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title