Learn More
IgG Mouse anti-Human, Alexa Fluor 405, Clone: 5A9cc, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP236511AF405
Catalog No. NBP236511AF
Add to cart
Description
Description
IgG Monoclonal antibody specifically detects Antigen in Human samples. It is validated for Enzyme Linked Immunosorbent Assay.
Specifications
Specifications
IgG
5A9cc
ELISA
Purified
IGHG1
IgG from human serum
Protein A purified
RUO
Store at 4C in the dark.
Monoclonal
Class-specific for human IgG. No allotype restriction. Relatively high affinity. No cross-reactivity with human IgA, IgM, IgE. MAb 5A9 is Fc-region specific.
ELISA
Alexa Fluor 405
50mM Sodium Borate with 0.05% Sodium Azide
immunoglobulin heavy constant gamma 1 (G1m marker)
Mouse
IgG2a
0.2 ml
Adaptive Immunity, Epitope Tags, Immunology
Primary
3500
Human