Learn More
IgG Mouse anti-Human, Clone: B33/20, Abnova™
Mouse Monoclonal Antibody
Manufacturer:  Abnova Corporation MAB14285
Catalog No. 89-034-367
Add to cart
Description
Description
Mouse monoclonal antibody raised against native human IgG.
  • High specificity
  • High reactivity
Specifications
Specifications
IgG
B33/20
Mouse monoclonal antibody raised against native human IgG.
In 10 mM PBS (0.05% BSA, 0.05% sodium azide).
Liquid
Mouse
IgG1, kappa
100 ug
Store at 4°C.
Monoclonal
Human
Flow Cytometry, Immunofluorescence, Immunohistochemistry (PFA fixed)
Unconjugated
Flow Cytometry (0.5-1 ug/106 cells in 0.1 mL) Immunofluorescence (0.5-1 ug/mL) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (0.5-1 ug/mL) The optimal working dilution should be determined by the end user.
P01857;P01859;P01860;P01861
IGHG1
Native purified polyclonal human Ig gamma chain.
Protein A/G purification
RUO
Primary
3500|3501|3502|3503
Documents
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Cancel Submit
For Research Use Only (RUO)