Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IgG, Rabbit, Polyclonal, Isotype Control, Novus Biologicals™
Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP2219520.25MG
Description
IgG Polyclonal specifically detects IgG in Rabbit samples. It is validated for .Specifications
IgG | |
Unconjugated | |
Rabbit | |
0.25 mg | |
Reconstitute with 1.0 mL PBS. | |
Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles. | |
IgG |
Polyclonal | |
0.2 um filtered solution in PBS. | |
Protein A or G purified | |
Isotype Control | |
Rabbit | |
Lyophilized |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title