Learn More
Medchemexpress LLC IgG Rabbit T185A 50ug
SDP
Supplier:  Medchemexpress LLC HYP7390450UG
Encompass_Preferred
IgG protein exhibits the D12 allotypic marker with the 104-Thr sequence and the E14 marker with 185-Thr IgG Protein Rabbit (T185A N284S HEK293) is the recombinant Rabbit-derived IgG protein expressed by HEK293 with tag free
Catalog No. 50-001-79410