Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IgG1 Armenian Hamster, BUV563, Clone: A19-3, Isotype Control, BD Horizon™
Armenian Hamster Monoclonal Antibody
Supplier: BD Biosciences 612933
Description
IgG1 Monoclonal is validated for Flow Cytometry, Control.Specifications
IgG1 | |
A19-3 | |
Brilliant Ultraviolet 563 | |
50 μg | |
Primary | |
IgG1 κ |
Flow Cytometry | |
0.2 mg/mL | |
Armenian Hamster | |
RUO | |
Refrigerate (2 to 8°C) |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction