Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IgG1 Human, HRP, Clone: , Isotype Control, Southern Biotech™

Human Isotype Control
Supplier: Southern Biotechnology Associates 0151K05
Description
SouthernBiotech isotype controls and purified immunoglobulins are useful as negative controls and standards in a variety of immunoassays.
Specifications
ELISA | |
HRP | |
Human | |
RUO | |
Store at 2-8°C |
Lot specific | |
Supplied in a stock solution in 50% glycerol/50% PBS, pH 7.4; No preservative added | |
1.0 mL | |
Primary | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction