Learn More
Enzo Life SciencesSupplier Diversity Partner Mouse IgG1 isotype control, monoclonal antibody (MOPC-21) (PE conjugate) (200 µg)  DFS Item
Manufacturer:  Enzo Life SciencesSupplier Diversity Partner ADISAB600PEF
Clone: MOPC-21. Host: Mouse. Isotype: IgG1. Source: Purified from ascites. Species Reactivity: Species independent. Applications: Flow Cytometry. Recommended dilutions: Flow Cytometry (10µg/ml). Suggested dilutions/conditions may not be available for all applications. Optimal conditions must be determined individually for each application. Purity: Protein G-affinity purified. Formulation: Liquid. In PBS, pH 7.2, containing 0.09% sodium azide. Handling: Avoid freeze/thaw cycles. Protect from light.
Catalog No. 50-667-395