Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse IgG1 kappa, Alexa Fluor 532, clone: P3.6.2.8.1, Isotype Control, eBioscience™ Non-distribution product as customer accommodation.

Mouse Isotype Control

Manufacturer:  Invitrogen 58471480

Catalog No. 50-112-4714


Add to cart

Description

Description

The isotype of a primary antibody and the application it is being used in can result in background staining. Primary antibody background noise can be caused by binding to Fc receptors on target cells; by non-specific interactions with cellular proteins, carbohydrates, and lipids; or by cell autofluorescence. Isotype control antibodies can act as negative controls to help differentiate non-specific background signal from specific antibody signal because they have no relevant specificity to a target antigen. While isotype controls are most commonly used in flow cytometry, they are useful in other applications such as chromatin immunoprecipitation (ChIP), immunohistochemistry, and gel shifts. Isotype controls should match with the primary antibody species and isotype so that the level of specific staining by the primary antibody may be accurately determined. If using directly labeled primary antibodies, the isotype control works best if conjugated with the same label as the test antibody.

Description: The monoclonal mouse IgG1 K immunoglobulin is useful as an isotype control.Applications Reported: This P3.6.2.1 antibody has been reported for use in flow cytometric analysis.Applications Tested: This P3.6.2.8.1 antibody has been tested by flow cytometric analysis of mouse splenocytes and human peripheral blood cells. This should be used at the same concentration as the experimental antibody.Alexa Fluor™ 532 is excited with the Green laser (532 nm) and emits at 561 nm. This cannot be used with the Yellow-Green laser (561 nm). We recommend using a 560/14 band pass filter. Please make sure that your instrument is capable of detecting this fluorochome.Excitation: 532 nm; Emission: 561 nm; Laser: Green Laser
Specifications

Specifications

Mouse IgG1 kappa
Control
0.2mg/mL
PBS with 0.1% gelatin and 0.09% sodium azide; pH 7.2
IgG1, IgG1k
IgG1, kappa
25μg
Store at 2-8°C. Do not freeze. Light-sensitive material.
Monoclonal
Control, Flow Cytometry
P3.6.2.8.1
Alexa Fluor 532
Liquid
Murine
Affinity chromatography
RUO
Isotype Control
Not Applicable
Documents
Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only.