Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse IgG1 kappa, eFluor 570, clone: P3.6.2.8.1, Isotype Control, eBioscience™ Non-distribution product as customer accommodation.

Mouse Isotype Control

Manufacturer:  Invitrogen 41471482

 View more versions of this product

Catalog No. 50-112-3617


Add to cart

Description

Description

The isotype of a primary antibody and the application it is being used in can result in background staining. Primary antibody background noise can be caused by binding to Fc receptors on target cells; by non-specific interactions with cellular proteins, carbohydrates, and lipids; or by cell autofluorescence. Isotype control antibodies can act as negative controls to help differentiate non-specific background signal from specific antibody signal because they have no relevant specificity to a target antigen. While isotype controls are most commonly used in flow cytometry, they are useful in other applications such as chromatin immunoprecipitation (ChIP), immunohistochemistry, and gel shifts. Isotype controls should match with the primary antibody species and isotype so that the level of specific staining by the primary antibody may be accurately determined. If using directly labeled primary antibodies, the isotype control works best if conjugated with the same label as the test antibody.

Description: The monoclonal mouse IgG1 K immunoglobulin is useful as an isotype control immunoglobulin.Applications Reported: This P3.6.2.8.1 antibody has been reported for use in immunohistochemical and immunocytochemical staining.Applications Tested: This Mouse IgG1 K Isotype Control eFluor™ 570 has been tested by immunocytochemistry on fixed and permeabilized MDCK cells and by immunohistochemistry (IHC-P) on formalin-fixed paraffin embedded human brain tissue. Use isotype control at the same concentration as experimental antibody.Filter Recommendation: When using this eFluor™ 570 antibody conjugate, we recommend a filter that will capture the 570 emission wavelength (for example, Excitation 545/25, 565LP, Emission 605/70). A standard Alexa Fluor™ 555 or TRITC filter is acceptable.Excitation: 555 nm; Emission: 570 nm
Specifications

Specifications

Mouse IgG1 kappa
Control
0.2mg/mL
PBS with 0.1% gelatin and 0.09% sodium azide; pH 7.2
IgG1, IgG1k
IgG1, kappa
100μg
Store at 2-8°C. Do not freeze. Light-sensitive material.
Monoclonal
Control, Immunocytochemistry, Immunofluorescence, Immunohistochemistry
P3.6.2.8.1
eFluor 570
Liquid
Murine
Affinity chromatography
RUO
Isotype Control
Not Applicable
Documents
Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only.