Learn More
Mouse IgG1 kappa Isotype Control (P3.6.2.8.1), eFluor™ 660, eBioscience™, Invitrogen™
Mouse Isotype Control
Supplier: Invitrogen 50471482
Description
Description: The monoclonal mouse IgG1 K immunoglobulin is useful as an isotype control. Applications Reported: The Mouse IgG1 K Isotype Control has been reported for use in flow cytometric analysis and immunohistochemical staining. Applications Tested: The Mouse IgG1 K Isotype Control has been tested by flow cytometric analysis of normal human peripheral blood cells. This can be used at the same concentration as the experimental antibody. eFluor® 660 is a replacement for Alexa Fluor® 647. eFluor® 660 emits at 659 nm and is excited with the red laser (633 nm). Please make sure that your instrument is capable of detecting this fluorochome. Excitation: 633-647 nm; Emission: 668 nm; Laser: Red Laser. Filtration: 0.2 μm post-manufacturing filtered.
Mouse IgG1 kappa Control antibody specifically detects Mouse IgG1 kappa in Not Applicable samples. It is validated for Control, Flow Cytometry, Immunohistochemistry.Specifications
Mouse IgG1 kappa | |
P3.6.2.8.1 | |
eFluor 660 | |
IgG1; Igh-4; Ighg1; Immunoglobulin G; Immunoglobulin G1; immunoglobulin heavy constant gamma 1 (G1m marker); ImmunoglobulinG; ImmunoglobulinG1; VH7183 | |
Affinity chromatography | |
RUO | |
Not Applicable | |
Liquid |
Control, Flow Cytometry, Immunohistochemistry | |
0.2 mg/mL | |
PBS with 0.09% sodium azide; pH 7.2 | |
Mouse | |
100 μg | |
Isotype Control (Primary) | |
4° C, store in dark, DO NOT FREEZE! | |
IgG1 κ |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
For Research Use Only.