Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rat IgG1 kappa, PE-Cyanine5, clone: eBRG1, Isotype Control, eBioscience™ Non-distribution product as customer accommodation.

Rat Isotype Control

Manufacturer:  Invitrogen 15430181

 View more versions of this product

Catalog No. 50-141-06


Add to cart

Description

Description

The isotype of a primary antibody and the application it is being used in can result in background staining. Primary antibody background noise can be caused by binding to Fc receptors on target cells; by non-specific interactions with cellular proteins, carbohydrates, and lipids; or by cell autofluorescence. Isotype control antibodies can act as negative controls to help differentiate non-specific background signal from specific antibody signal because they have no relevant specificity to a target antigen. While isotype controls are most commonly used in flow cytometry, they are useful in other applications such as chromatin immunoprecipitation (ChIP), immunohistochemistry, and gel shifts. Isotype controls should match with the primary antibody species and isotype so that the level of specific staining by the primary antibody may be accurately determined. If using directly labeled primary antibodies, the isotype control works best if conjugated with the same label as the test antibody.

Description: The monoclonal rat IgG1, kappa is useful as an isotype control immunoglobulin.Applications Reported: PE-Cy5 Rat IgG1 Iso Cntrl has been reported for use in flow cytometric analysis.Applications Tested: This PE-Cy5 Rat IgG1 isotype control antibody should be used at the same concentration as the experimental antibody.Excitation: 488-561 nm; Emission: 667 nm; Laser: Blue Laser, Green Laser, Yellow-Green Laser.Filtration: 0.2µm post-manufacturing filtered.
Specifications

Specifications

Rat IgG1 kappa
Control
0.2mg/mL
PBS with 0.1% gelatin and 0.09% sodium azide; pH 7.2
IgG1, IgG1k
IgG1, kappa
50μg
Store at 2-8°C. Do not freeze. Light-sensitive material.
Monoclonal
Control, Flow Cytometry
eBRG1
PE-Cyanine5
Liquid
Rat
Affinity chromatography
RUO
Isotype Control
Not Applicable
SDS
Documents
Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only.