Learn More
IgG1 Kpa ItCl Rat, R718, Clone: R3-34, Isotype Control, BD Horizon™
Rat Monoclonal Antibody
Manufacturer:  BD Biosciences 566948
Catalog No. BDB566948
Add to cart
Description
Description
Following immunization of a rat with mouse immunoglobulin (Ig), the Ig from the R3-34 hybridoma was identified as a non-reactive clone. The R3-34 immunoglobulin was selected as an Ig isotype control following screening for low background staining on a variety of mouse and human cells and tissues.
Specifications
Specifications
IgG1 Kpa ItCl
R3-34
Liquid
50 μg
Refrigerate Upon Arrival
Monoclonal
Flow Cytometry
R718
Rat
RUO
Isotype Control