Learn More
IgG1 Kpa ItCl Rat, R718, Clone: X40, Isotype Control, BD Horizon™
Rat Monoclonal Antibody
Manufacturer:  BD Biosciences 566928
Catalog No. BDB566928
Add to cart
Description
Description
The X40 hybridoma was derived from the fusion of mouse Sp2/0-Ag 14 myeloma cells with spleen cells from BALB/c mice immunized with keyhole limpet hemocyanin (KLH). The hybridoma secretes monoclonal antibody that binds specifically to KLH, an antigen not expressed on human cells or human cell lines.
Specifications
Specifications
IgG1 Kpa ItCl
X40
Liquid
50 μg
Refrigerate Upon Arrival
Monoclonal
Flow Cytometry
R718
Rat
RUO
Isotype Control