Learn More
IgG1, lambda 1 Hamster, Brilliant Ultraviolet 737, Clone: G235-2356, Isotype Control, BD Horizon™
Hamster Isotype Control
Manufacturer:  BD Biosciences 612820
Catalog No. BDB612820
Add to cart
Description
Description
IgG1, lambda 1 Monoclonal isotype control is validated for Flow cytometry.
Specifications
Specifications
IgG1, lambda 1
G235-2356
Brilliant Ultraviolet 737
Affinity Purified
Trinitrophenol-KLH
Affinity Chromatography
RUO
Monoclonal
Flow Cytometry
0.2mg/mL
Aqueous buffered solution containing ≤0.09% sodium azide.
Hamster
IgG1, λ 1
50 μg
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.