Learn More
IgG1 Mouse, BUV563, Clone: X40, Isotype Control, BD Horizon™
Mouse Monoclonal Antibody
Manufacturer:  BD Biosciences 612920
Catalog No. BDB612920
Add to cart
Description
Description
IgG1 Monoclonal is validated for Flow Cytometry, Control.
Specifications
Specifications
IgG1
X40
Brilliant Ultraviolet 563
Mouse
50 μg
Refrigerate (2 to 8°C)
Flow Cytometry, Control
0.2 mg/mL
Conjugated, Purified
IgG1, κ