Learn More
Enzo Life SciencesSupplier Diversity Partner IgG1 (mouse), ELISA kit (1x96wells)  DFS Item
Manufacturer:  Enzo Life SciencesSupplier Diversity Partner ADI900109
The IgG1 (mouse), EIA kit is a colorimetric immunometric enzyme immunoassay kit with results in 90 minute. Absorbance is read at 450 nm. Ideal for determining the isotype and concentration of monoclonal antibodies. Sensitivity: 0.064 ng/mL (range 7.81 - 250 ng/ml). Assay Time: 90 minutes. Applications: ELISA, Colorimetric detection. Species Reactivity: Mouse. Contents: GxM IgG Microtiter plate, mouse IgG1 conjugate, Assay buffer 13 concentrate, Wash buffer concentrate, mouse IgG1 standard, TMB Substrate, Stop solution 2
Catalog No. 50-658-147