Learn More
IgG1 Rat anti-Mouse, Brilliant Violet 510, Clone: X56, BD Optibuild™
Rat Monoclonal Antibody
Manufacturer:  BD Biosciences 742476
Catalog No. BDB742476
Add to cart
Description
Description
The X56 monoclonal antibody specifically binds to mouse IgG1 of Igh-C[a] /and Igh-C[b] /haplotypes. It does not crossreact with other Ig isotypes.
The antibody was conjugated to BD Horizon™ BV510 which is part of the BD Horizon Brilliant™ Violet family of dyes. With an Ex Max of 405nm and Em Max at 510nm, BD Horizon BV510 can be excited by the violet laser and detected in the BD Horizon V500 (525/50nm) filter set. BD Horizon BV510 conjugates are useful for the detection of dim markers off the violet laser. BV510 is a polymer-based dye that is brighter than BD Horizon V500. Due to similar excitation and emission properties, BD Horizon BV510 and BD Horizon V500 cannot be used simultaneously.
Specifications
Specifications
IgG1
0.2mg/mL
Aqueous buffered solution containing ≤0.09% sodium azide.
Igh-4, Ighg1, Immunoglobulin heavy constant gamma 1
Mouse IgG1, κ Antibody
The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated with BD Horizon BV510 under optimal conditions that minimize unconjugated dye and antibody.
RUO
Primary
Murine
X56
Brilliant Violet 510
Affinity Purified
Rat
Rat LOU, also known as Louvain, LOU/C, LOU/M IgG1, κ
50μg
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Monoclonal