Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IgG1, κ Rat, Brilliant Violet 750, Clone: R3-34, Isotype Control BD

Rat Monoclonal Antibody

Manufacturer:  BD Biosciences 566367

Catalog No. BDB566367


Add to cart

Description

Description

Following immunization of a rat with mouse immunoglobulin (Ig), the Ig from the R3-34 hybridoma was identified as a non-reactive clone. The R3-34 immunoglobulin was selected as an Ig isotype control following screening for low background staining on a variety of mouse and human cells and tissues.

The antibody was conjugated to BD Horizon BV750 which is part of the BD Horizon Brilliant™ Violet family of dyes. This dye is a tandem fluorochrome of BD Horizon BV421 with an Ex Max of 405nm and an acceptor dye with an Em Max at 750nm. BD Horizon Brilliant BV750 can be excited by the violet laser (405 nm) and detected with a 750/30 nm filter with a 740 nm long pass. Due to spectral differences between labeled cells and beads, using BD™ CompBeads can result in incorrect spillover values when used with BD Horizon BV750 reagents. Therefore, the use of BD CompBeads or BD CompBeads Plus to determine spillover values for these reagents is not recommended. BV750
Specifications

Specifications

Rat IgG1, κ Isotype Control
0.2mg/mL
Aqueous buffered solution containing ≤0.09% sodium azide.
Rat
Rat IgG1, κ
RUO
Primary
R3-34
Brilliant Violet 750
IgG1 kappa Isotype Control
Mouse immunoglobulin
50μg
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Monoclonal
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit