Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IgG1, γ, Rat, BV480, Clone: A110-1; Isotype Control, BD Horizon™
Rat Monoclonal Isotype Control
Supplier: BD Biosciences 566128
Description
IgG1, γ Monoclonal antibody specifically detects IgG1, γ in samples. It is validated for Isotype control, Flow cytometry.Specifications
IgG1 γ | |
Monoclonal | |
0.2mg/mL | |
Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
Trinitrophenyl–Keyhole Limpet Hemocyanin | |
RUO | |
Primary | |
Rat LOU, also known as Louvain, LOU/M IgG1 γ |
Flow Cytometry | |
A110-1 | |
Brilliant Violet 480 | |
Rat | |
50 μg | |
Cell Biology | |
Purified |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Reseach Use Only.