Learn More
IgG2, λ1 Hamster, Brilliant Ultraviolet 737, Clone: Ha4/8, Isotype Control, BD Horizon™
Hamster Isotype Control
Manufacturer:  BD Biosciences 612822
Catalog No. BDB612822
Add to cart
Description
Description
IgG2, λ1 Monoclonal isotype control is validated for Flow cytometry.
Specifications
Specifications
IgG2, λ1
Ha4/8
Brilliant Ultraviolet 737
Affinity purified
Keyhole limpet hemocyanin
Affinity Chromatography
RUO
Monoclonal
Flow Cytometry
0.2mg/mL
Aqueous buffered solution containing ≤0.09% sodium azide.
Hamster
IgG2, λ1
50 μg
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.