Learn More
IgG2 Armenian Hamster, BV750, Clone: B81-3, Isotype Control, BD Horizon™
Armenian Hamster Isotype Control
Manufacturer:  BD Biosciences 566370
Catalog No. BDB566370
Add to cart
Description
Description
IgG2 antibody specifically detects IgG2 in samples. It is validated for Flow Cytometry.
Specifications
Specifications
IgG2
B81-3
Brilliant Violet 750
Armenian Hamster
50 μg
Refrigerate (2 to 8°C)
Flow Cytometry
0.2 mg/mL
Purified
IgG2, κ