Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IgG2 Rabbit anti-Human, Biotin, Clone: RM118, Secondary Antibody, Novus Biologicals™

Rabbit Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP262076B

Catalog No. NBP262076B


Add to cart

Description

Description

IgG2 Monoclonal antibody specifically detects IgG2 in Human samples. It is validated for Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry.
Specifications

Specifications

IgG2
RM118
Biotin
50% Glycerol/PBS with 1% BSA with 0.09% Sodium Azide
Constant region of heavy chain of IgG2, immunoglobulin gamma 2 (Gm marker), immunoglobulin heavy constant gamma 2 (Gm marker)
Rabbit
IgG
50 ug
Secondary
3501
Human
ELISA, Flow Cytometry, Immunocytochemistry, Immunohistochemistry
1 mg/mL
Flow Cytometry, ELISA .05ug/mL - 0.2ug/mL, Immunohistochemistry 0.5ug/mL - 2ug/mL, Immunocytochemistry 0.5ug/mL - 2ug/mL
Purified
IGHG2
Human IgG2
Protein A purified
Store at -20°C. Avoid freeze-thaw cycles.
Monoclonal
This antibody reacts to the heavy chain of human IgG2. This antibody does not cross react to any other IgG subclasses (IgG1, IgG3, or IgG4), and shows no cross reactivity to IgM, IgA, IgD, or IgE. It does not react to monkey (Cyno or Rhesus) IgG, mouse IgG, rat IgG, or goat IgG.
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit