Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IgG2 Rabbit anti-Human, Clone: RM118, Abnova™

Rabbit monoclonal antibody raised against native human IgG2.

Manufacturer:  Abnova Corporation MAB12819

Catalog No. 89-934-328


Add to cart

Description

Description

Specifications

Specifications

IgG2
RM118
Rabbit monoclonal antibody raised against native human IgG2.
In PBS (50% glycerol, 1% BSA, 0.09% sodium azide)
DKFZp686I04196
Rabbit
IgG
100 ug
Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Monoclonal
This antibody reacts to the heavy chain of human IgG2. No cross reactivity with human IgG1, IgG3, IgG4, IgM, IgA, IgD, IgE, mouse IgG, rat IgG, or goat IgG.
ELISA, Flow Cytometry, Immunocytochemistry, Immunohistochemistry
Unconjugated
ELISA (0.05-0.2 ug/mL for detection; 0.05-0.2 ug/well for capture) Flow Cytometry Immunocytochemistry (0.5-2 ug/mL) Immunohistochemistry (0.5-2 ug/mL) The optimal working dilution should be determined by the end user.
Liquid
IGHG2
Native purified human IgG2.
Protein A affinity purification
RUO
Primary
3501
Human
SDS
Documents
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit