Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IgG2a, Brilliant Violet 786, Clone: MOPC-173, Isotype Control, BD

Manufacturer:  BD Biosciences 565032

Catalog No. BDB565032


Add to cart

Specifications

Specifications

IgG2a
MOPC-173
Brilliant Violet 786
Affinity purified
None
50μg
Secondary
Flow Cytometry
0.2mg/mL
Aqueous buffered solution containing BSA and ≤0.09% sodium azide
Murine
IgG2a, κ
4°C, protect from prolonged exposure to light. Do not freeze.
Monoclonal
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit